Learn More
Electron Microscopy Sciences INFINITYX 2, 8, 18 and 32 Megapixel Pixel Shifting USB 2.0 Microscopy Camera, CMOS, Color
SDP
Catalog No. 5032319
Encompass
Using patented pixel shifting technology, each CCD pixel captures full chromatic information resulting in increased color resolution with no degradation. The INFINITYX-32 is the first pixel shifting camera to utilize the highly sensitive SONY ICS274 sensor. This complete imaging solution is an affordable, extraordinary tool for professionals in clinical, life and material sciences where megapixel resolution, color accuracy and light sensitivity are essential.
Sensor: CCD
Type: Color
Catalog No. 50-323-19 Supplier Electron Microscopy Sciences Supplier No. 95116
Add to Cart
Add to Cart
Request a Quote
Using patented pixel shifting technology, each CCD pixel captures full chromatic information resulting in increased color resolution with no degradation. The INFINITYX-32 is the first pixel shifting camera to utilize the highly sensitive SONY ICS274 sensor. This complete imaging solution is an affordable, extraordinary tool for professionals in clinical, life and material sciences where megapixel resolution, color accuracy and light sensitivity are essential.
Sensor: CCD
Type: Color