Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ INFINITY 8.9 MP CMOS Monochrome Camera
GREENER_CHOICESDP

Catalog No. 01189186 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Pixels:
8.9 MP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pixels
01-189-186 8.9 MP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-186 Supplier ACCU-SCOPE™ Supplier No. I8LM0009M
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The INFINITY 8-9M delivers exceptional monochrome images with ultra-high definition with superb low light performance. Included is the latest microscopy software, INFINITY ANALYZE 7. This software suite provides a breadth of features including support for both brightfield and fluorescence imaging.

  • Monochrome CMOS sensor with 1.0 in. optical format
  • Global shutter
  • 2.9 megapixel camera with fast frame rates of 62 fps at full resolution
  • Direct USB3 interface for fast image delivery and simplified connectivity
  • Compact form factor
  • Support for locking industrial micro USB on the back for power and control
  • INFINITY ANALYZE microscopy software included with purchase. No annual license fees. (Camera requires INFINITY ANALYZE 7.0.3 or higher)
  • Software support for Windows and Mac, along with third-party plugins for ImagePro, MetaMorph and MicroManager
  • Selectable 8 or 12-bit pixel data
  • Recommended coupler: 1.0x

Specifications

Type USB Microscope Camera
Autoclavable Not Autoclavable
Pixels 8.9 MP
Sterility Non-sterile
Voltage 5 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.