Learn More
Electron Microscopy Sciences INFINITY1 1.3 Megapixel USB 2.0 Microscopy Camera, CMOS, Monochrome
SDP
Catalog No. 5032310
Encompass
INFINITY1 CMOS USB 2.0 digital cameras are designed to be a cost-effective, versatile solution for clinical, life science, materials science and educational professionals. Features include auto white balance, full exposure control, programmable gain, sub-windowing, and region of interest. It is an excellent fit for documentation and archiving applications.
Features 2, 3, and 5 mexapixel resolution. It is perfect for documentation and archiving applications and has fast frame rates.
Sensor: CMOS
Type: Monochrome
Catalog No. 50-323-10 Supplier Electron Microscopy Sciences Supplier No. 95100
INFINITY1 CMOS USB 2.0 digital cameras are designed to be a cost-effective, versatile solution for clinical, life science, materials science and educational professionals. Features include auto white balance, full exposure control, programmable gain, sub-windowing, and region of interest. It is an excellent fit for documentation and archiving applications.
Features 2, 3, and 5 mexapixel resolution. It is perfect for documentation and archiving applications and has fast frame rates.
Sensor: CMOS
Type: Monochrome