Learn More
Electron Microscopy Sciences INFINITY 3S-1UR Monochrome Camera CCD
SDP
Catalog No. 501928126
Encompass_Preferred
The INFINITY3S-1UR is a high-speed, ultra-sensitive research grade microscopy camera with 1.4 megapixel resolution. The camera incorporates Sony's new ICX825 CCD sensor, producing much higher dynamic range and sensitivity over the popular ICX285 sensor. The INFINITY3S-1UR has the unmatched light sensitivity needed for challenging low light applications, such as fluorescence and NIR imaging. Lumenera's high-speed USB 3.0 data interface provides maximum allowable throughput where full resolution images can be sent to a host computer at an industry leading 60 fps.
Catalog No. 50-192-8126 Supplier Electron Microscopy Sciences Supplier No. 95132
Add to Cart
Add to Cart
Request a Quote
The INFINITY3S-1UR is a high-speed, ultra-sensitive research grade microscopy camera with 1.4 megapixel resolution. The camera incorporates Sony's new ICX825 CCD sensor, producing much higher dynamic range and sensitivity over the popular ICX285 sensor. The INFINITY3S-1UR has the unmatched light sensitivity needed for challenging low light applications, such as fluorescence and NIR imaging. Lumenera's high-speed USB 3.0 data interface provides maximum allowable throughput where full resolution images can be sent to a host computer at an industry leading 60 fps.