Learn More
Laxco Infinity Plan Achromatic Objective 50X with Logo.oil Immersion
GREENER_CHOICESDP
Catalog No. 12563729 Shop All Laxco Inc Products
Change view
Click to view available options
Magnification Power:
50X
Catalog No. Magnification Power
12-563-729 50X
1 options
Catalog No. 12-563-729 Supplier Laxco Supplier No. LMCPOIP50 OIL
Add to Cart
Add to Cart
Research grade optical quality enables optimum clarity and reproducibility. Objective is spring loaded to ensure slide safty and smooth transition in between oil immersion objectives. Suitable for hematology. Works best with LAXCO LMC Series upright microscopes.
  • Infinity achromatic infinity corrected 50X oil objective
  • Numerical aperture A0.95
  • Working Distance 0.17 mm
  • Research grade optical quality
  • Ideal for hematology applications
  • Works best with LAXCO LMC series upright microscopes
Specifications
Type Objective
Autoclavable Not Autoclavable
Magnification Power 50X
Sterility Sterile