Learn More
Agilent Technologies InfinityLab Long-Life Deuterium DAD/MWD Lamp, RFID Tag, 1 ea
Catalog No. NC1759191
Encompass
This long-life deuterium lamp features an RFID tag and is designed for use with specific diode array detectors and multiple wavelength detectors in high-performance liquid chromatography systems. Each unit is manufactured in an ISO 9001 certified environment, providing full traceability and consistent performance through rigorous testing against optical standards.
  • Guaranteed to last longer than 2,000 hours
  • Higher initial intensity from 190 nm to 330 nm
  • Superior signal stability with reduced stabilization time
  • Lower limits of detection and decreased noise levels
  • Best lamp-to-lamp consistency for reliable results
Catalog No. NC1759191 Supplier Agilent Technologies Supplier No. 21400820
This long-life deuterium lamp features an RFID tag and is designed for use with specific diode array detectors and multiple wavelength detectors in high-performance liquid chromatography systems. Each unit is manufactured in an ISO 9001 certified environment, providing full traceability and consistent performance through rigorous testing against optical standards.
  • Guaranteed to last longer than 2,000 hours
  • Higher initial intensity from 190 nm to 330 nm
  • Superior signal stability with reduced stabilization time
  • Lower limits of detection and decreased noise levels
  • Best lamp-to-lamp consistency for reliable results