Learn More
Agilent Technologies InfinityLab Quick Turn Capillary, Stainless Steel, 0.12 mm ID, 105 mm Length, 1 ea
Catalog No. NC3800864
Encompass
This stainless steel capillary is designed for use in analytical liquid chromatography applications. With an inner diameter of 0.12 mm and a length of 105 mm, this component is intended to be combined with compatible Quick Turn LC fittings for secure, low-dispersion fluidic connections within InfinityLab LC instruments.
  • Manufactured from durable stainless steel
  • Compatible with InfinityLab Quick Turn fitting (5067-5966)
  • Features an inner diameter of 0.12 mm and a length of 105 mm
  • Designed for analytical LC system configurations
  • Rated for high-pressure operations up to 1,300 bar
  • Supports up to 200 reconnections
Catalog No. NC3800864 Supplier Agilent Technologies Supplier No. 55001188
This stainless steel capillary is designed for use in analytical liquid chromatography applications. With an inner diameter of 0.12 mm and a length of 105 mm, this component is intended to be combined with compatible Quick Turn LC fittings for secure, low-dispersion fluidic connections within InfinityLab LC instruments.
  • Manufactured from durable stainless steel
  • Compatible with InfinityLab Quick Turn fitting (5067-5966)
  • Features an inner diameter of 0.12 mm and a length of 105 mm
  • Designed for analytical LC system configurations
  • Rated for high-pressure operations up to 1,300 bar
  • Supports up to 200 reconnections