Learn More
Agilent Technologies INFINITYLAB STAY SAFE CAP KIT
Catalog No. NC2211153
Encompass
InfinityLab stay safe cap, GL45, 4 ports for waste. Includes 4 fittings (2 x 3.2 mm, 1 x 2.5 mm, 1 x 1.6 mm) and 1 leak hose
This item is not returnable. View return policy
Catalog No. NC2211153 Supplier Agilent Technologies Supplier No. 50431220
InfinityLab stay safe cap, GL45, 4 ports for waste. Includes 4 fittings (2 x 3.2 mm, 1 x 2.5 mm, 1 x 1.6 mm) and 1 leak hose