Learn More
Neta Scientific INFINITYLAB THERMAL EQUILIBRAT
SDP
Catalog No. NC1999997
Encompass_Preferred
This item is manufactured by Agilent
  • Supports diverse analytical methods.
  • Accelerates thermal equilibration of columns.
  • Ensures consistent column temperatures for reproducible results.
  • Accommodates both short and long chromatography columns.
  • Designed for use with specific compatible systems.
  • Installation in top row of Multicolumn Thermostat.
  • Includes silicone side covers and twist-lock clips.
Catalog No. NC1999997 Supplier Neta Scientific Supplier No. G711660013
This item is manufactured by Agilent
  • Supports diverse analytical methods.
  • Accelerates thermal equilibration of columns.
  • Ensures consistent column temperatures for reproducible results.
  • Accommodates both short and long chromatography columns.
  • Designed for use with specific compatible systems.
  • Installation in top row of Multicolumn Thermostat.
  • Includes silicone side covers and twist-lock clips.