Learn More
Cayman Chemical Inflachromene, Purity: ≥95%, CAS: 908568-01-4, 10 mg
SDP
Catalog No. 502966083
Encompass_Preferred
An anti-inflammatory agent that directly binds HMGB1 and HMGB2 and reduces their cytoplasmic accumulation in microglial cells; downregulates proinflammatory functions of HMGB proteins and reduces neuronal damage in vivo; ameliorates inflammatory pathogenesis in a mouse model of sepsis
Catalog No. 50-296-6083 Supplier Cayman Chemical Supplier No. 1700610 MG
An anti-inflammatory agent that directly binds HMGB1 and HMGB2 and reduces their cytoplasmic accumulation in microglial cells; downregulates proinflammatory functions of HMGB proteins and reduces neuronal damage in vivo; ameliorates inflammatory pathogenesis in a mouse model of sepsis