Learn More
Cayman Chemical Inflammasome Inhibitor 4b, Purity: ≥98%, CAS: 2768759-64-2, 1 mg
SDP
Catalog No. 502914954
Encompass_Preferred
An inhibitor of NLRP3 inflammasome activation; inhibits ATP-induced IL-1β release by 98.3% in LPS-primed mouse BMDMs at 1 µM; decreases paw swelling and mechanical hyperalgesia in a mouse model of gout induced by monosodium urate crystals at 3 and 10 mg/kg
Catalog No. 50-291-4954 Supplier Cayman Chemical Supplier No. 370301 MG
An inhibitor of NLRP3 inflammasome activation; inhibits ATP-induced IL-1β release by 98.3% in LPS-primed mouse BMDMs at 1 µM; decreases paw swelling and mechanical hyperalgesia in a mouse model of gout induced by monosodium urate crystals at 3 and 10 mg/kg