Learn More
Raybiotech Inc INFLAMMATION ARRAY C3 (8)
Catalog No. 501043154
Encompass_Preferred
RayBio C-Series Human Inflammation Antibody Array 3 Kit. Detects 40 Human Inflammatory Factors. Suitable for all liquid sample types.
Catalog No. 50-104-3154 Supplier Raybiotech Inc Supplier No. AAHINF38
RayBio C-Series Human Inflammation Antibody Array 3 Kit. Detects 40 Human Inflammatory Factors. Suitable for all liquid sample types.