Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Vein Thrombus Control Slides
SDP

Catalog No. 21010275 Shop All Mopec Products
Change view
Click to view available options
Quantity:
100 Control Slides
25 Control Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-010-275 100 Control Slides
21-010-276 25 Control Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-010-275 Supplier Mopec Supplier No. SC958100
Only null left
Add to Cart
Add to Cart

Normal Human Tissue Vein Thrombus Control Slides

  • High-quality control slides to support accurate histopathology and laboratory testing
  • Includes well-preserved normal human vein tissue for reliable comparison and analysis
  • Easy-to-use slides suitable for both laboratory and educational purposes
  • Prepared to maintain tissue integrity for consistent experimental and diagnostic results

Specifications

Content And Storage Room-Temperature
For Use With (Application) Use as a Control
Type Control Slide
Quantity 100 Control Slides
Includes 98 Unstained 2 Stained Control Slides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.