Learn More
Fabohio Inc INFLATABLE FRAME GLOVEBAG
SDP
Catalog No. NC1626034
Encompass
Inflatable frame glovebag having beveled front similar to display model expect size: 38” wide x 23.5” x 26” high add 2 barbed hose fittings for Nitrogen purge Material of Construction: Polyurethane
Catalog No. NC1626034 Supplier Fabohio Inc Supplier No. 20MNJ100238
Inflatable frame glovebag having beveled front similar to display model expect size: 38” wide x 23.5” x 26” high add 2 barbed hose fittings for Nitrogen purge Material of Construction: Polyurethane