Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Replogle Inflatable Solar System
SCI_ED

Catalog No. S81627
Change view
Click to view available options
Grade:
K+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81627 K+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81627 Supplier Replogle Supplier No. 17801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Explore the wonders of astronomy with Replogle's Inflatable Solar System Set, and turn any space into outerspace.

  • Inflate a planet and easily transform any setting into a personal planetarium with this fun, decorative and educational set.
  • The color and design of this set is based on actual representations of the planets' view from space including cloud cover and weather systems.
  • The set includes a large 28 in. diameter Sun along with Mercury, Venus, Earth, Mars, Jupiter, Saturn,Uranus, Neptune, and Pluto which are all proportional in size.
  • Each planet is made of easy-to-clean and durable vinyl and features built-in mounting loops so you can easily string your set indoors or outdoors (mounting string not included).

Specifications

Description Inflatable Solar System
Subject Astronomy
Grade K+
For Use With (Application) Instructional Learning
Includes Sun, Mercury, Venus, Earth, Mars, Jupiter, Saturn, Uranus, Neptune, Pluto
Diameter (English) 8 to 28 in.
Quantity 1 Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.