Learn More
Grainger Inflating Gauge w/CH-330LO Chuck
Catalog No. 502750650
Encompass_Preferred
The Grainger 140Z25 is a 25-foot, 14 AWG, 3-conductor SJOOW portable power cable. It has a yellow jacket and is rated at 600 volts. The cable has stranded copper conductors with EPDM insulation. It is flexible, oil resistant, and designed for use outdoors or in wet areas. The 140Z25 can be used to provide power to lighting, tools, motors and other equipment in industrial, construction and maintenance applications. With proper connections and circuit protection, it allows users to safely run power over longer distances.
Catalog No. 50-275-0650 Supplier Grainger Supplier No. 140Z25
The Grainger 140Z25 is a 25-foot, 14 AWG, 3-conductor SJOOW portable power cable. It has a yellow jacket and is rated at 600 volts. The cable has stranded copper conductors with EPDM insulation. It is flexible, oil resistant, and designed for use outdoors or in wet areas. The 140Z25 can be used to provide power to lighting, tools, motors and other equipment in industrial, construction and maintenance applications. With proper connections and circuit protection, it allows users to safely run power over longer distances.