Learn More
Med Vet International McKesson LUMEON Blood Pressure Bulb and Cuff Nylon Medium Cuff Black 23 - 40 cm Arm Adult 01-865-11ABKGM 1/Box
Catalog No. 503073795
Encompass_Preferred
McKesson LUMEON Blood Pressure Bulb and Cuff Nylon Medium Cuff Black 23 - 40 cm Arm Adult 01-865-11ABKGM 1/Box
Catalog No. 50-307-3795 Supplier Med Vet International Supplier No. 803206BX
McKesson LUMEON Blood Pressure Bulb and Cuff Nylon Medium Cuff Black 23 - 40 cm Arm Adult 01-865-11ABKGM 1/Box