Learn More
Medchemexpress LLC HY-P9970 1mg Medchemexpress, Infliximab CAS:170277-31-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99701MG
Encompass_Preferred
Medchemexpress, HY-P9970 1mg Infliximab CAS:170277-31-3 Infliximab (Avakine) is a chimeric monoclonal IgG1 antibody that specifically binds to TNF-α. Infliximab prevents the interaction of TNF-α with TNF-α receptor (TNFR1 and TNFR2). Infliximab has the potential for autoimmune, chronic inflammatory diseases and diabetic neuropathy research[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1777