Learn More
Abcam Infliximab ELISA Kit, 96TEST
Supplier:  Abcam AB23764796TEST
Encompass_Preferred
Infliximab ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Infliximab in Human in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5181