Learn More
Advanced ImmunoChemical, Inc. Influenza A (H3N2) Virus (Strain A/Shangdong/9/93), 0.05mg
Supplier:  Advanced ImmunoChemical, Inc. 7IVAH3N20.1MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Influenza A (H3N2) virus (strain A/Shangdong/9/93). Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Shangdong/9/93. Applications: Serological studies of influenza A virus, immunogen for antibody production. Analysis: Purity > 90 % Tested with anti-influenza A monoclonal antibodies in ELISA. Purification: Ultracentrifugation with 10 – 40 % sucrose gradient. Presentation: STE, 0.09 % sodium azide (NaN3) and 0.005 % thimerosal. Storage Conditions: -20 °C (between -15 and -30 °C allowed). Inactivation: Thimerosal and beta propiolactone treatment. This product has been treated in a manner consistent with methods of inactivation. Generally accepted good laboratory practices appropriate to microbiological/viral safe handling practices and techniques are required at work. Material Safety Note: This product is sold for research use only.
Catalog No. 50-153-9860