Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Influenza A H1N1 2009 (Swine Flu) Culture Fluid
SDP

Catalog No. 22157670 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Strain Designations:
Canada Isolate
NY Isolate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Strain Designations
22-157-670 NY Isolate
22-157-669 Canada Isolate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-670 Supplier ZeptoMetrix Supplier No. 0810109CFN
Add to Cart
Add to Cart

IMPORTANT: Contains live material, so cust must submit an executed ZeptoMetrix MTA before vendor can ship. Also, a USDA APHIS VS 16-6 or 16-6A authorized permit must be obtained and a valid copy of the permit received by ZeptoMetrix prior to shipment of this product. requests.

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Content And Storage -65°C
Form Solution
Media Type Vial
Product Type Influenza A H1N1 Virus Infectious Culture Fluid
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Strain Designations NY Isolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.