Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA-specific B cell probe (Y108F, His & AVI Tag), Biotinylated(HPLC-verified)
Supplier:  Sino Biological 11055V49BB20
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/California/04/2009) (H1N1)) hemagglutinin (translated amino acids of ACP41105.1) (Y108F, Met1-Gly519), termed as HA-specific B cell probe, was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. NC2306386