Learn More
Sino Biological Influenza A H1N1 (A/Arizona/13/2008) Neuraminidase / NA (His Tag) 100ug
Supplier:  Sino Biological 40734V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Arizona/13/2008 (H1N1)) neuraminidase (ACM51249) (His36-Lys470), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-211-5852
 Restricted Item