Learn More
Sino Biological Influenza A H1N1 (A/Brisbane/02/2018) Hemagglutinin / HA Protein (His Tag) 100ug
Supplier:  Sino Biological 40719V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Brisbane/02/2018 (H1N1)) hemagglutinin (translated amino acids of EPI1312566) (Met1-Ile530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5839