Learn More
Sino Biological Influenza A H1N1 (A/Brisbane/02/2018) Neuraminidase / NA Protein (ECD, His Tag) 100ug
Supplier:  Sino Biological 40767V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Brisbane/02/2018 (H1N1)) neuraminidase (translated amino acids of EPI1799927) (Ile30-Lys469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6019
 Restricted Item