Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA Protein (His Tag), Biotinylated
Supplier:  Sino Biological 11055V08BB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/California/04/2009 (H1N1)) (ACP41105.1) hemagglutinin (Met1-Gln529) (Native, HA1+HA2, uncleaved) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5418