Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 11058V07B100
Encompass_Preferred
A DNA sequence encoding the mature form of influenza A virus (A/California/04/2009 (H1N1)) neuraminidase (ACP41107.1) (His 36-Lys 469) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-211-5429