Learn More
Sino Biological Influenza A H1N1 (A/California/07/2009) Hemagglutinin / HA Protein (His Tag), Biotinylated
Supplier:  Sino Biological 11085V08HB20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus H1N1 (A/California/07/2009) (ACP41953.1) hemagglutinin (Met1-Gln529) (HA1+HA2, uncleaved) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5419