Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Neuraminidase / NA (Active)
Supplier:  Sino Biological 11058VNAHC200
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/California/04/2009 (H1N1)) neuraminidase (ACP41107.1) (His 36-Lys 469) was expressed, the cell lysates are collected, and bio-activity was tested.
Catalog No. 50-224-6803