Learn More
Sino Biological Influenza A H1N1 (A/Guangdong-Maonan/SWL1536/2019) / (A/Hawaii/70/2019) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40717V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Guangdong-Maonan/SWL1536/2019) / (A/Hawaii/70/2019) (H1N1)) hemagglutinin (translated amino acids of EPI1542570 & QHN72768.1) (Met1-Ile530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5838