Learn More
Sino Biological Influenza A H1N1 (A/Michigan/45/2015) Nucleoprotein / NP Protein (His Tag) 100ug
Supplier:  Sino Biological 40777V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Michigan/45/2015 (H1N1)) nucleoprotein (translated amino acids of EPI685572) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4267
 Restricted Item