Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/1934) Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11684T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/Puerto Rico/8/1934) Hemagglutinin / HA ( Catalog#11684-V08H; ABD77675.1; Met1-Gln528). Influenza A H1N1 (A/Puerto Rico/8/1934) Hemagglutinin / HA specific IgG was purified by Influenza A H1N1 (A/Puerto Rico/8/1934) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-224-7498