Learn More
Sino Biological Influenza A H1N1 (A/swine/Canada/01093/2006) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40733V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A H1N1 (A/swine/Canada/01093/2006) Neuraminidase (AKM14549) (Met1-Gln430) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and polyhistidine tag at the C-terminus.
Catalog No. 50-224-6500