Learn More
Sino Biological Influenza A H1N1 (A/Victoria/2454/2019) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40851V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A H1N1 (A/Victoria/2454/2019) Nucleoprotein (EPI1840657) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6507