Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/28/2011) Hemagglutinin / HA Protein (His Tag) 100ug
Supplier:  Sino Biological 40675V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Wisconsin/28/2011 (H1N1)) hemagglutinin (AEX38474) (Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5797