Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/588/2019) / (A/Victoria/2570/2019) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40787V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/588/2019) / (A/Victoria/2570/2019) (H1N1)) hemagglutinin (translated amino acids of EPI1661758 & EPI1799581) (Met1-Ile530), termed as HA, was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4234