Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/588/2019) / (A/Victoria/2570/2019) Neuraminidase / NA Protein (His Tag)
Supplier:  Sino Biological 40785V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/588/2019) / (A/Victoria/2570/2019) (H1N1)) neuraminidase (translated amino acids of EPI1661757 & EPI1799580) (Ile30-Lys469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4233
 Restricted Item