Learn More
Sino Biological Influenza A H1N1 Hemagglutinin / HA Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11684MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant H1N1 (A/Puerto Rico/8/1934) HA (Catalog#11684-V08H; ABD77675.1; Met 1-Gln 528). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7504