Learn More
Creative Biomart Subunit Influenza virus Vaccine Antigen
Catalog No. 502084701
Encompass_Preferred
Recombinant Full-Length H1N1 New Caledonia/20/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is approximately 72 kDa.The insect cells infected with A9440.1a recombinant baculovirus expressing recombinant H1N1 A/New Caledonia/20/99.H1N1 New Caledonia shows 90% similarity to the A/PR/8/34 amino acid sequence.The accession number is DQ508857.
Catalog No. 50-208-4701 Supplier Creative Biomart Supplier No. H1N1323
Recombinant Full-Length H1N1 New Caledonia/20/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is approximately 72 kDa.The insect cells infected with A9440.1a recombinant baculovirus expressing recombinant H1N1 A/New Caledonia/20/99.H1N1 New Caledonia shows 90% similarity to the A/PR/8/34 amino acid sequence.The accession number is DQ508857.