Learn More
Sino Biological Influenza A H1N1 (Swine Flu 2009) Neuraminidase / NA Antibody, Mouse MAb
Supplier:  Sino Biological 11058MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant influenza A virus H1N1 Neuraminidase. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. NC1501796