Learn More
Sino Biological Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 ELISA Pair Set 5 Plates
Catalog No. 502221498
Encompass_Preferred
The Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 ELISA Pair Set is for the quantitative determination of Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 coated on a 96-well plate. Standards and samples are added to the wells, and any Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Immunodeficiency Virus type 1 (HIV-1)
Catalog No. 50-222-1498 Supplier Sino Biological Supplier No. SEK116955
The Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 ELISA Pair Set is for the quantitative determination of Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 coated on a 96-well plate. Standards and samples are added to the wells, and any Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Immunodeficiency Virus type 1 (HIV-1)