Learn More
ZeptoMetrix Influenza A H1N1 (Wisconsin/67/22) Virus Culture Fluid
SDP
Catalog No. 22032258 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-032-258 1 mL
1 options
Catalog No. 22-032-258 Supplier ZeptoMetrix Supplier No. 0810685CF
Add to Cart
Add to Cart
Viral culture fluids are sold as consumable testing materials, and are not for propagation or commercialization.
  • This product is intended for research, analytical testing during product development, or quality assurance testing
  • Applications include nucleic acid-molecular testing, limit of detection (LOD) studies, cross-reactivity studies, and other viral-based assays
Specifications
Product Type Infectious Culture Fluid
Sterility Non-sterile
Quantity 1 mL