Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Influenza A H1N1pdm (New York/18/09) Culture Fluid
SDP

Catalog No. 22158127 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-127 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-127 Supplier ZeptoMetrix Supplier No. 0810519CF
Add to Cart
Add to Cart

For research, product development testing, or quality assurance testing

Influenza virus is an enveloped virus with a diameter of 80-120nm and contains a single-stranded, segmented, negative-sense RNA within a nucleocapsid.

  • Propagated in the MDCK cell line
  • Each lot of viral culture fluid is assayed for its Tissue Culture Infective Dose (TCID50) and sold with titers >105U/mL

Specifications

Biosafety Level 2
Content And Storage 2°C to 8°C
For Use With (Application) Nucleic acid/molecular testing, Limit of Detection (LOD) studies, Cross-reactivity studies, Other viral-based assays
Quantity 1 mL

Do not use in humans. For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.