Learn More
Sino Biological Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin / HA1 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11707T46100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin / HA1 ( Catalog#11707-V08H1; AAA43178.1; Met1-Arg345). Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin / HA1 specific IgG was purified by Influenza A H3N2 (A/Aichi/2/1968) Hemagglutinin / HA1 affinity chromatography.
Catalog No. 50-228-3165