Learn More
Sino Biological Influenza A H3N2 (A/Cambodia/e0826360/2020) Hemagglutinin / HA Protein (His Tag) 100ug
Supplier:  Sino Biological 40789V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Cambodia/e0826360/2020 (H3N2)) hemagglutinin (translated amino acid of EPI1837753) (Met1-Asp529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4235