Learn More
Sino Biological Influenza A H3N2 (A/Cambodia/e0826360/2020) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40778V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Cambodia/e0826360/2020 (H3N2)) nucleoprotein (translated amino acids of EPI1841678) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4268
 Restricted Item