Learn More
Sino Biological Influenza A H3N2 (A/Darwin/9/2021) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40859V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Darwin/9/2021) Hemagglutinin (EPI1859996) (Met1-Asp529) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6513