Learn More
Sino Biological Influenza A H3N2 (A/Darwin/9/2021) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40860V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Darwin/9/2021) Neuraminidase (EPI1859995) (Thr30-Ile469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6514