Learn More
Sino Biological Influenza A H3N2 (A/Hong Kong/2671/2019) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40721V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/2671/2019 (H3N2)) hemagglutinin (translated amino acids of EPI1698489) (Met1-Asp529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5841