Learn More
Sino Biological Influenza A H3N2 (A/Hong Kong/2671/2019) Neuraminidase / NA Protein (His Tag)
Supplier:  Sino Biological 40847V07H100
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Hong Kong/2671/2019) Neuraminidase (EPI1698488) (Thr31-Thr469) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6504